Skip to Content
Merck
All Photos(1)

Key Documents

CLS480137

Corning® LSE rotors for LSE compact centrifuges

fixed angle rotor for 12 x 15 mL tubes, 6,000rpm, 1/cs

Synonym(s):

fixed angle centrifuge rotor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909
NACRES:
NB.15

feature

to hold 12 x 15 mL tubes

packaging

case of 1
pack of 1

manufacturer/tradename

Corning 480137

parameter

200-6000 max. RCF

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models LSE Compact

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service