Recommended Products
Product Name
AtT-20/D16v-F2, 94050406
biological source
mouse brain
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse pituitary tumour
Cell Line Description
This F2 subclone was developed from original AtT-20, an andrenocorticotropic hormone (ACTH) secreting cell line established from murine pituitary tumour. These cells are readily transfected using a standard calcium phosphate protocol.
Application
AtT-20/D16v-F2 has been used to study the role of the G-protein Rasd1 in hypothalamic response to neuronal activation. It has also been used to study the effect of cAMP and glucocorticoid treatment on cAMP-responsive element binding protein 3 like 1 (CREB3L1).
Culture Medium
DMEM + 2mM Glutamine + 10% FBS.
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:5 using 0.25% trypsin/EDTA; 5% CO2; 37°C. Cell clumps must be broken up to prevent cells growing in suspension.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service