Skip to Content
Merck
All Photos(1)

Documents

CCD-39Lu

90110512, human lung, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

CCD-39Lu, 90110512

biological source

human lung

growth mode

Adherent

karyotype

Modal no. 46

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian lung

Cell Line Description

Derived from a lung explant of a Caucasian female infant with mildly severe hyaline membrane disease and hypoxia.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10,12
D13S317: 11
D16S539: 11,13
D5S818: 11,12
D7S820: 9,12
THO1: 6,9.3
TPOX: 8,11
vWA: 14,17

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:3 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service