Skip to Content
Merck
All Photos(1)

Key Documents

MM386 cell line human

10092318, human skin, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

MM386 cell line human, 10092318

biological source

human skin

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human malignant melanoma

Cell Line Description

Malignant Melanoma. Cell Bank Australia reference number: CBA1349.

DNA Profile

Not specified

Culture Medium

RPMI 1640 (with 2 mM L-Glutamine + 25 mM HEPES) +10% FBS.

Subculture Routine

Optimal split ratio 1:4 (seeding density 1.2 x104 cells/cm2).Cells incubated at 37 °C with 5% CO2.Harvest the cells using 0.05% Trypsin/EDTA at 37 °C for 5 min.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments.  Please check with your local Customer Service representative for more information.
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Depositor and originator: Peter Parsons - Queensland Institute of Medical Research, Australia

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service