C9275
Calcium Ionophore A23187 hemicalcium salt
powder
Synonym(s):
A23187 hemicalcium salt, Antibiotic A 23187 hemicalcium salt, Calcimycin hemicalcium salt, Calimycin hemicalcium salt
About This Item
sterility
non-sterile
Quality Level
form
powder
color
yellow
solubility
DMSO: soluble
ethanol: soluble
antibiotic activity spectrum
Gram-positive bacteria
fungi
Mode of action
cell membrane | interferes
storage temp.
2-8°C
SMILES string
CNc1ccc2oc(C[C@H]3O[C@@]4(CC[C@H]3C)O[C@@H]([C@H](C)C[C@H]4C)[C@H](C)C(=O)c5ccc[nH]5)nc2c1C(=O)O[Ca]OC(=O)c6c(NC)ccc7oc(C[C@H]8O[C@@]9(CC[C@H]8C)O[C@@H]([C@H](C)C[C@H]9C)[C@H](C)C(=O)c%10ccc[nH]%10)nc67
InChI
1S/2C29H37N3O6.Ca/c2*1-15-10-11-29(17(3)13-16(2)27(38-29)18(4)26(33)20-7-6-12-31-20)37-22(15)14-23-32-25-21(36-23)9-8-19(30-5)24(25)28(34)35;/h2*6-9,12,15-18,22,27,30-31H,10-11,13-14H2,1-5H3,(H,34,35);/q;;+2/p-2/t15-,16-,17-,18-,22-,27+,29?;15-,16-,17-,18-,22-,27+,29+;/m11./s1
InChI key
LAQWEYASNNRUGY-PLYBNSMSSA-L
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service