Skip to Content
Merck
All Photos(1)

Documents

B6936

Sigma-Aldrich

3-Benzidino-6-(4-chlorophenyl)pyridazine

≥98% (HPLC), solid

Synonym(s):

BCP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H17N4Cl
CAS Number:
Molecular Weight:
372.85
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

light yellow

solubility

DMSO: >14 mg/mL

storage temp.

2-8°C

SMILES string

Nc1ccc(cc1)-c2ccc(Nc3ccc(nn3)-c4ccc(Cl)cc4)cc2

InChI

1S/C22H17ClN4/c23-18-7-1-17(2-8-18)21-13-14-22(27-26-21)25-20-11-5-16(6-12-20)15-3-9-19(24)10-4-15/h1-14H,24H2,(H,25,27)

InChI key

ABGCSSPCKSVMHI-UHFFFAOYSA-N

Biochem/physiol Actions

3-Benzidino-6-(4-chlorophenyl)pyridazine is an inhibitor of delayed rectifier and transient outward potassium currents. The IC50 values for the blocking action of BCP on IKDR and IKA was calculated as 7.13 μM and 0.55 μM, respectively in acutely isolated rat hippocampal pyramidal neurons by using whole-cell patch-clamp technique. The parent compound, minaprine (Cat. No. M3157), has selective affinity for M1 muscarinic receptors and possesses memory-enhancing properties and also acts as an antidepressant.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura F Grice et al.
Molecular biology and evolution, 34(5), 1083-1099 (2017-01-21)
Although discriminating self from nonself is a cardinal animal trait, metazoan allorecognition genes do not appear to be homologous. Here, we characterize the Aggregation Factor (AF) gene family, which encodes putative allorecognition factors in the demosponge Amphimedon queenslandica, and trace
Thomas R Schulz et al.
Journal of medical virology, 90(2), 271-276 (2017-09-09)
Hepatitis B virus (HBV) from 76 adult immigrants in Australia from Myanmar was characterized to determine the prevalence of different HBV genotypes and subgenotypes. A mutational analysis was then performed to determine the presence of clinically significant mutations and correlate
D Wong et al.
Alimentary pharmacology & therapeutics, 47(1), 114-122 (2017-10-13)
Hepatitis B e antigen (HBeAg) seroconversion is a treatment endpoint for HBeAg-positive CHB, and a necessary precursor to HBsAg loss. Biomarkers that predict serological outcomes would be useful. To evaluate the utility of measuring HBeAg levels for predicting HBeAg seroconversion
Joseph D Hill et al.
Scientific reports, 7(1), 5250-5250 (2017-07-14)
Bulk fabrication of surface patterns with sub-20 nm feature sizes is immensely desirable for many existing and emerging technologies. Directed self-assembly (DSA) of block copolymers (BCPs) has been a recently demonstrated approach to achieve such feature resolution over large-scale areas with
Anne-Els van de Logt et al.
Kidney international, 95(6), 1514-1517 (2019-05-06)
Differences between laboratory assays can have important clinical implications. For creatinine assays this became apparent soon after the introduction of the Modification of Diet in Renal Disease formula and resulted in international efforts towards standardization. Albumin in blood is measured

Articles

We offer many products related to potassium channels for your research needs.

We offer many products related to potassium channels for your research needs.

We offer many products related to potassium channels for your research needs.

We offer many products related to potassium channels for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service