Skip to Content
Merck
All Photos(1)

Key Documents

02128

Sigma-Aldrich

Zinc bromide

puriss., anhydrous, ≥98%

Synonym(s):

Zinc dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnBr2
CAS Number:
Molecular Weight:
225.20
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38150410
PubChem Substance ID:

grade

puriss.

Assay

≥98%

reaction suitability

reagent type: catalyst
core: zinc

mp

394 °C (lit.)

anion traces

sulfate (SO42-): ≤100 mg/kg

cation traces

Ba: ≤10 mg/kg
Ca: ≤100 mg/kg
Fe: ≤10 mg/kg
K: ≤100 mg/kg
Mg: ≤100 mg/kg
Na: ≤100 mg/kg
Pb: ≤50 mg/kg

SMILES string

Br[Zn]Br

InChI

1S/2BrH.Zn/h2*1H;/q;;+2/p-2

InChI key

VNDYJBBGRKZCSX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Zinc bromide is an inorganic salt. Zinc bromide supported on hydroxyapatite (Zn-HAP) promotes the synthesis of 3-substituted indoles.

Application

Zinc bromide (ZnBr2) may be used for the following studies:
  • Removal of dimethoxytrityl ethers from deoxynucleosides.
  • Removal of N-acyl protecting group in nucleoside derivatives.
  • Alkylation of O-silylated dienolates of unsaturated ketones and esters.
  • As catalyst in the synthesis of α-perfluoroalkylated homoallylic alcohols.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The use of zinc bromide for removal of dimethoxytrityl ethers from deoxynucleosides.
Matteucci MD and Caruthers MH.
Tetrahedron Letters, 31(34), 3243-3246 (1984)
Zinc bromide supported on hydroxyapatite as a new and efficient solid catalyst for Michael addition of indoles to electron-deficient olefins.
Tahir R, et al.
J. Mol. Catal. A: Chem., 246(1), 39-42 (2006)
Zinc bromide promoted allylation of aluminum acetals derived from perfluoro carboxylic acid esters and diisobutylaluminum hydride. New convenient access to a-perfluoroalkylated homoallyl alcohols.
Ishihara T, et al.
Tetrahedron Letters, 34(36), 5777-5777 (1993)
γ-Alkylation of unsaturated ketones and esters: zinc bromide-catalysed alkylation of O-silylated dienolates.
Fleming I, et al.
Tetrahedron Letters, 20(34), 3209-3212 (1979)
Yejian Han et al.
Organic letters, 13(4), 588-591 (2011-01-12)
An efficient and versatile ZnBr(2)-catalyzed Diels-Alder/carbocyclization cascade reaction has been developed for construction of highly functionalized cis-hydrindanes (70-96% yields with high diastereoselectivity), and it has been successfully utilized in the synthesis of the aglycon of dendronobiloside A.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service