Skip to Content
Merck

CCW950

Rose Bengal Chloramphenicol Agar Plates

plate diam. 90 mm, ready-to-use, sterile, box of 20 plates, for yeasts, for molds

Synonym(s):

RBCA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.74
UNSPSC Code:
41171606
Composition:
agar, 15.0 g/L , chloramphenicol, 0.10 g/L , D(+)-glucose, 10.0 g/L , magnesium sulfate, 0.50 g/L , mycological peptone, 5.00 g/L , potassium phosphate, 1.00 g/L , rose bengal, 0.05 g/L
Plate diam.:
90 mm
Feature:
ready-to-use
Sterility:
sterile
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

ISO 11133

Quality Segment

sterility

sterile

form

solid

shelf life

3.5 mo. at 2‑14 (Expiry date on the label)

feature

ready-to-use

composition

agar, 15.0 g/L , chloramphenicol, 0.10 g/L , D(+)-glucose, 10.0 g/L , magnesium sulfate, 0.50 g/L , mycological peptone, 5.00 g/L , potassium phosphate, 1.00 g/L , rose bengal, 0.05 g/L

packaging

box of 20 plates

storage condition

(Store dry and tightly closed)

parameter

21 mL filling volume

plate diam.

90 mm

color

strongly pink

pH

7.2±0.2 (25 °C)

application(s)

food and beverages
microbiology

storage temp.

2-8°C

suitability

molds, yeasts

General description

Rose Bengal Chloramphenicol Agar is a selective medium used to detect and enumerate molds and yeasts in food samples. In addition to the nutritional requirements for molds and yeasts, this medium also contains Rose Bengal, which turns the yeast a pink color and facilitates counting by reducing the luxuriant growth of molds such as Rhizopus and Neurospora. This makes it easier to detect other slower-growing molds. The chloramphenicol included in the formulation inhibits bacterial growth but does not interfere with the growth of fungi.

Application

Rose Bengal Chloramphenicol Agar Plates are ready-to-use plates suitable for the isolation of yeast and molds from the environment and food products.

Other Notes

  • 1 box with 2 packs of 10 plates/pack
  • Single cellophane


Storage Class

13 - Non Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library