Skip to Content
Merck

78080BC

MVP ICON® USB Cable

for use with MVP ICON® instrument

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116500
NACRES:
NA.28
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

MVP ICON®

manufacturer/tradename

MVP ICON®

application(s)

food and beverages

compatibility

for use with MVP ICON® instrument

storage temp.

room temp

Legal Information

MVP ICON is a registered trademark of Merck KGaA, Darmstadt, Germany




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library