Skip to Content
Merck
All Photos(1)

Key Documents

32211-M

Sigma-Aldrich

Chloroform

puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)

Synonym(s):

Methylidyne trichloride, Trichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
Beilstein:
1731042
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.21
technique(s):
RNA extraction: suitable
bp:
60.5-61.5 °C (lit.)
vapor pressure:
160 mmHg ( 20 °C)

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor density

4.1 (vs air)

vapor pressure

160 mmHg ( 20 °C)

grade

puriss. p.a.

Assay

99.0-99.4% (GC)

form

liquid

contains

~1% ethanol as stabilizer

technique(s)

RNA extraction: suitable

impurities

≤0.00001% free chlorine (Cl)
≤0.00005% free acid (as HCl)
≤0.0005% non-volatile matter
≤0.005% aldehydes and ketones (as CH3COCH3)
≤0.005% carbonyl compounds (as CO)
≤0.01% tetrachloroethene (GC)
≤0.01% tetrachloromethane (GC)
≤0.01% trichloroethene (GC)
≤0.01% water (Karl Fischer)
≤0.03% dichloromethane (GC)
0.6-1.0% ethanol (GC)

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

density

1.476-1.483 g/mL at 20 °C
1.492 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.0001%

cation traces

Al: ≤0.50 ppm
B: ≤0.02 ppm
Ba: ≤0.10 ppm
Ca: ≤0.50 ppm
Cd: ≤0.05 ppm
Co: ≤0.02 ppm
Cr: ≤0.02 ppm
Cu: ≤0.02 ppm
Fe: ≤0.10 ppm
Mg: ≤0.10 ppm
Mn: ≤0.02 ppm
Ni: ≤0.02 ppm
Pb: ≤0.05 ppm
Sn: ≤0.10 ppm
Zn: ≤0.10 ppm

SMILES string

ClC(Cl)Cl

suitability

complies for appearance
complies for reaction against H2SO4
complies for suitability of determ. w. dithizone

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloroform, a halogenated hydrocarbon, is widely employed as a solvent. Its photochemical degradation has been investigated in the presence of TiO2 aqueous suspensions (wavelength range of 310-380nm).

Application

Chloroform may be employed as a solvent for the following studies:
  • Extraction of RNA from plant cells.
  • Preparation of dioleoylphosphatidylcholine (DOPC) solution.
  • Dissolution of 1,2-distearoyl-sn-glycero-3-phosphocholine (DSPC).
It may be employed in a rapid quantitative method for the release of periplasmic proteins from bacterial strains.
Chloroform may be used as a solvent to enhance the enantioselectivity of secondary alcohols formed by the borohydride reduction of carbonyl compounds in the presence of optically active ketoiminatocobalt complexes.

Other Notes

For information on chloroform miscibility, please visit the following link:
Chloroform Miscibility/Immiscibility Table

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gurman Singh Pall et al.
Nucleic acids research, 35(8), e60-e60 (2007-04-05)
The northern blot, or RNA gel blot, is a widely used method for the discovery, validation and expression analysis of small regulatory RNA such as small interfering RNA (siRNA), microRNA (miRNA) and piwi-interacting RNA (piRNA). Although it is straightforward and
Z V Leonenko et al.
Biochimica et biophysica acta, 1509(1-2), 131-147 (2000-12-19)
We have used magnetic alternating current mode atomic force microscopy (MAC-AFM) to investigate the formation of supported phospholipid bilayers (SPB) by the method of vesicle fusion. The systems studied were dioleoylphosphatidylcholine (DOPC) on mica and mica modified with 3-aminopropyl-triethoxy-silane (APTES)
Jameel A Feshitan et al.
Journal of colloid and interface science, 329(2), 316-324 (2008-10-28)
Microbubbles used as contrast agents for ultrasound imaging, vectors for targeted drug delivery and vehicles for metabolic gas transport require better size control for improved performance. Mechanical agitation is the only method currently available to produce microbubbles in sufficient yields
G F Ames et al.
Journal of bacteriology, 160(3), 1181-1183 (1984-12-01)
We introduce a method by which periplasmic proteins can be released rapidly, simply, and quantitatively by treating cells with chloroform. All the amino acid-binding proteins tested maintained their activity during chloroform treatment. This method makes practical the analysis of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service