Skip to Content
Merck
All Photos(1)

Documents

16682

Sigma-Aldrich

(R)-Pantoic acid sodium salt

≥95% (GC)

Synonym(s):

(2R)-2,4-Dihydroxy-3,3-dimethylbutanoic acid monosodium salt, (R)-2,4-Dihydroxy-3,3-dimethylbutyric acid sodium salt, Sodium (R)-pantoate, Sodium D-pantoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NaO4
CAS Number:
Molecular Weight:
170.14
Beilstein:
6119584
UNSPSC Code:
12352106
NACRES:
NA.25

Quality Level

Assay

≥95% (GC)

optical activity

[α]/D 8.0±1.0°, c = 1 in H2O

storage temp.

−20°C

InChI

1S/C6H12O4.Na/c1-6(2,3-7)4(8)5(9)10;/h4,7-8H,3H2,1-2H3,(H,9,10);/q;+1/p-1/t4-;/m0./s1

InChI key

XTIMSSMEPPKSQD-WCCKRBBISA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

(R)-Pantoic acid is an intermediate in the phosphopantothenate biosynthesis pathway for the de novo synthesis of R-pantothenate (vitamin B5) from valine in plants and microorganisms.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

13C-NMR and 1H-1H NOEs of Coenzyme A: conformation of the pantoic acid moiety.
Dordine, R.L., et al.
Bioorganic Chemistry, 23, 169-181 (1995)
Analysis of stereochemistry of enzymically formed pantoyl lactone or pantoic acid by gas chromatography and circular dichroism.
D R Wilken et al.
Analytical biochemistry, 112(1), 9-16 (1981-03-15)
Michael Rothmann et al.
Journal of the American Chemical Society, 135(16), 5962-5965 (2013-04-05)
Pantetheine and its corresponding disulfide pantethine play a key role in metabolism as building blocks of coenzyme A (CoA), an essential cofactor utilized in ~4% of primary metabolism and central to fatty acid, polyketide, and nonribosomal peptide synthases. Using a
Hiroya Tomita et al.
Journal of bacteriology, 194(19), 5434-5443 (2012-08-07)
Although bacteria and eukaryotes share a pathway for coenzyme A (CoA) biosynthesis, we previously clarified that most archaea utilize a distinct pathway for the conversion of pantoate to 4'-phosphopantothenate. Whereas bacteria/eukaryotes use pantothenate synthetase and pantothenate kinase (PanK), the hyperthermophilic
Dayong Si et al.
Applied microbiology and biotechnology, 93(4), 1619-1625 (2011-11-16)
Ketopantoic acid (KPA) reductase catalyzes the stereospecific reduction of ketopantoic acid to D-pantoic acid. Based on the N-terminal amino acid sequence of KPA reductase from Stenotrophomonas maltophilia 845, the KPA reductase gene was cloned from S. maltophilia NBRC14161 and sequenced.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service