Skip to Content
Merck

ZFCBIQXUS

Country Box Milli-Q® IQ Systems

Available only in USA, Canada and Puerto Rico

Synonym(s):

Country Box Milli-Q® IQ 7003/05/10/15 US

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
JA.13
UNSPSC Code:
41104214
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 1 kit

availability

available only in USA, Canada and Puerto Rico

compatibility

for use with Milli-Q® IQ 7003, for use with Milli-Q® IQ 7005, for use with Milli-Q® IQ 7010, for use with Milli-Q® IQ 7015

General description

The Country Box for Milli-Q® IQ 7003/05/10/15 is an installation accessory that contains a quick reference guide, a user manual in local language on a USB key, and a power cord adapted to the local electrical network.

Application

The components of the country box assist users to quickly access the information they need to correctly use their system and connect to the local power network.

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Country Box Milli-Q® IQ Systems




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library