Skip to Content
Merck
All Photos(2)

Documents

P3B050A00

Millipore

Pellicon® 3 Cassette with Biomax® Membrane

50kDa, A Screen, 88 cm2

Synonym(s):

Pellicon® 3 Cassette with Biomax® 50 kDa Membrane, A Screen, 88cm2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151806
eCl@ss:
32031623
NACRES:
NB.24

material

TPE seal
polyethersulfone membrane (void free)
polypropylene
polypropylene cap (for end)
polypropylene screen

Quality Level

feature

holdup volume 1.8 mL (in feed channel)

manufacturer/tradename

Pellicon®

parameter

30 psi max. reverse press. exposure at 25 °C (for 24 hours)
4-6 L/min-m2 recirculation rate
40 psi max. transmembrane pressure at 4-50 °C
5.5 bar max. inlet pressure (80 psi) at 30 °C
5.5 bar max. transmembrane pressure (80 psig) at 30 °C
6.9 bar max. inlet pressure (100 psi)
80 psi max. transmembrane pressure at 4-40 °C

technique(s)

analytical sample preparation: suitable
buffer exchange: suitable
protein purification: suitable (concentration)

H

0.8 cm (0.33 in.)

L

20.6 cm (8.1 in.)

W

5.6 cm (2.2 in.)

filtration area

88 cm2

membrane area

88 cm2

gravimetric extractables

≤1500 mg/m2

matrix

Biomax® (Membrane)

pore size

50 kDa NMWCO

pH

1-14

shipped in

ambient

General description

Pellicon® 3 filtration devices are advanced, high-performance cassettes ideal for today’s higher titer therapeutic antibodies as well as the more demanding filtration processes that require higher operating pressures, temperatures and caustic cleaning regimes.

The Pellicon® 3 Cassette with Biomax® Membrane is perfect for the filtration of monoclonal antibodies and solutions containing albumin, hormones, vaccines and growth factors.

This P3B050A00 Pellicon® 3 ultrafiltration device features a pore size of 50 kDa NMWCO, a filtration area of 88 cm2, a Biomax® 50 membrane, and dimensions measuring 0.8 cm (H) x 5.6 cm (W) x 20.6 cm (L).

Features and Benefits

  • Rugged & reliable – ideally suited for today′s more demanding filtration processes
  • High performance – produces excellent resolution, yields and back-pressure resistance
  • Optimum product recovery – thanks to proven membrane technology
  • Void-free technology – leading-edge membranes resolve virtually any separation challenge
  • Defect-free, superior adhesion – Ultracel®uses composite membranes cast on a microporous substrate
  • Automated manufacturing – delivers unbeatable performance, consistency and reliability
  • Easy installation & care – simple to install and maintain

Other Notes

These products are manufactured in a facility which adheres to FDA Good Manufacturing Practices.

Analysis Note

Gravimetric Extractables: after 24 hours in water at controlled room temperature

Legal Information

BIOMAX is a registered trademark of Merck KGaA, Darmstadt, Germany
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service