Skip to Content
Merck

MABD79

Anti-FOXG1 Antibody, clone 3G11.2

clone 3G11.2, from mouse

Synonym(s):

Forkhead box protein G1, Brain factor 1, BF-1, BF1, Brain factor 2, BF-2, BF2, hBF-2, Forkhead box protein G1A, Forkhead box protein G1B, Forkhead box protein G1C, Forkhead-related protein FKHL1, HFK1, Forkhead-related protein FKHL2, HFK2, Forkhead-relat

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
3G11.2, monoclonal
Application:
WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3G11.2, monoclonal

species reactivity

mouse, rat, human

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FOXG1(2290)

General description

Forkhead box protein G1 (FOXG1) is also called Brain factor 1 (BF-1), Brain factor 2 (BF-2), Forkhead-related protein FKHL1 (HFK1), Forkhead-related protein FKHL2 (HFK2) and Forkhead-related protein FKHL3 (HFK3). FOXG1 is a repressive transcription factor involved in establishing the regional subdivision of the developing brain and in the development of the telencephalon. FOXG1 expression is limited to the neurons of the developing telencephalon and is involved in Rett syndrome congenital variant (RTTCV).
~52 kDa observed

Immunogen

Linear peptide corresponding to human FOXG1.

Application

Detect FOXG1 using this mouse monoclonal antibody, Anti-FOXG1 Antibody & clone 3G11.2 validated for use in western blotting.
Research Category
Stem Cell Research
Research Sub Category
Neural Stem Cells
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected FOXG1 in 10 µg in human cortex and rat brain tissue lysate.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected FOXG1 in 10 µg of mouse brain tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library