IPAKICPK1
Milli-Q® IQ Element Consumable Kit
Removes trace ionic contaminants and particulates from ultrapure water
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104212
NACRES:
JA.13
Recommended Products
Quality Level
packaging
pkg of 1 kit
manufacturer/tradename
IPAK Quanta®
parameter
(Resistivity 18.2 MΩ.cm @ 25 °C)
1.5 L/min flow rate (Resistivity 18.2 MΩ.cm @ 25 °C)
total organic carbon (TOC) residue
≤5 ppb
pore size
0.1 μm filter (final filter)
compatibility
for use with Milli-Q® Element
Related Categories
General description
Milli-Q® IQ Element consumable kit consists of IPAK Quanta® ICP polishing cartridge and 0.1 μm Optimizer LW final filter. The polishing cartridge removes trace ions while the filter removes particulates from feed water to produce high-quality ultrapure water.
Application
Product ultrapure water is suitable for sensitive analyses by ICP-MS, GF-AAS and trace IC.
Features and Benefits
- For optimal purification of ultrapure water down to ppt and sub-ppt ion levels.
- Purification cartridges have been designed to be effortlessly replaced.
Legal Information
IPAK QUANTA is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service