EZFITL0HE1
Microfil® V or Microfil® S Filtration Head
available as 1 unit
Synonym(s):
For use with EZ-Fit® vacuum manifold, Microfil® V or Microfil® S filtration head for EZ-Fit® Manifold (1 unit)
About This Item
Recommended Products
material
stainless steel
Sterilization Compatibility
autoclavable compatible
manufacturer/tradename
EZ-Fit®
input
sample type cosmetics
sample type pharmaceutical(s)
sample type beer
sample type bottled water
application(s)
bioburden testing
cosmetics
environmental
food and beverages
water monitoring
shipped in
ambient
General description
Application
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service