Skip to Content
Merck

ABS528

Anti-S1P lyase Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

S1PL, SPL1, mSPL, Sphingosine-1-phosphate aldolase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
3
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Sgpl1(20397)

General description

S1P lyase is an important enzyme in lipid, steroid, and ceramide metabolism. S1P lyase 1 specifically cleaves phosphorylated sphingoid bases such as sphingosine-1-phosphate into fatty aldehydes and phosphoethanolamine. S1P lyase also plays roles in apoptosis with regards to regulation of ceramide production and plays roles in cell differentiation via its role in androgen and estrogen metabolism as well as that of other important lipids and bioactive lipid derivatives. S1P lyase is expressed in the endoplasmic reticulum in many cells. Interestingly, because Sphingosine-1-phosphate (S1P) is a bioactive sphingolipid that promotes cardiomyocyte survival and contributes to ischemic preconditioning, its catabolism by S1P lyase is very important in recovery from heart attacks or other ischemia related events, and thus S1P lyase is being examined as a potential new therapeutic target for reperfusion therapy in heart disease.
~ 56 kDa observed

Immunogen

Epitope: cytoplasmic domain
Linear peptide corresponding to the cytoplasmic domain of Mouse S1P lyase.

Application

Research Category
Signaling

Metabolism
Research Sub Category
Lipid Metabolism & Weight Regulation

Lipid Signaling
This Anti-S1P lyase Antibody is validated for use in Western Blotting for the detection of S1P lyase.
Western Blotting Analysis: A representative lot of this antibody detected S1P lyase in 30 μg of lysate from various mice tissues (Bektas, M., et al. (2010). Journal of Biological Chemistry Vol. 285, NO. 14, pp. 10880–10889)

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in mouse lung tissue lysate.

Western Blotting Analysis: 1 μg/mL of this antibody detected SP-lyase1 in 10 µg of mouse lung tissue lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Explore all of our products under Anti-S1P lyase Antibody

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library