ABS2090
Anti-ICDH
serum, from rabbit
Synonym(s):
Isocitrate dehydrogenase, ICDH (NADP), IDH
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
biological source
rabbit
Quality Level
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
E. coli, Legionella pneumophila
packaging
antibody small pack of 25 μL
technique(s)
western blot: suitable
isotype
IgG
GenBank accession no.
shipped in
ambient
target post-translational modification
unmodified
General description
Isocitrate dehydrogenase [NADP] (also known as IDH, ICDH) catalyzes the oxidative decarboxylation of isocitrate to alpha-ketoglutarate. In eukaryotes there are at least three isozymes of IDH: two are located in the mitochondrial matrix (one NAD+-dependent, the other NADP+-dependent), while the third one (also NADP+-dependent) is cytoplasmic. In E.coli the activity of a NADP+-dependent ICDH is controlled by the phosphorylation of a serine residue and the phosphorylated form of ICDH is completely inactivated. It has been reported that if Ser113 is replaced by alanine, cysteine, threonine, or tyrosine there is some loss of enzymatic activity. However, substitution by aspartic acid completely abolishes its activity. ICDH contains three domains: a large alpha + beta domain, a small alpha/beta domain and an alpha/beta clasp-like domain involving both subunits. (Ref.: Hurley, JH et al (1989). Proc. Natl. Acad. Sci. USA. 86 (22); 8635-8639).
Specificity
Expected to react with most eubacteria.
This polyclonal antibody detects isocitrate dehydrogenase in E. coli and Legionella pneumophila.
Immunogen
Epitope: unknown
His-tagged full length recombinant isocitrate dehydrogenase from Legionella pneumophila, strain Lp02.
Application
Anti-ICDH, Cat. No. ABS2090, is a rabbit polyclonal antibody that detects Isocitrate dehydrogenase [NADP]. It has been tested for use in Western Blotting.
Research Category
Signaling
Signaling
Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected ICDH in 20 µL Legionella cell lysate.
Western Blotting Analysis: A representative lot detected ICDH in E. Coli cell lysate (Xu, L., et. al. (2010). PLoS Pathog. 6(3):e1000822; Lu, C., et. al. (2016). Sci Signal. 9(412):ra11).
Western Blotting Analysis: A representative lot detected ICDH in E. Coli cell lysate (Xu, L., et. al. (2010). PLoS Pathog. 6(3):e1000822; Lu, C., et. al. (2016). Sci Signal. 9(412):ra11).
Quality
Evaluated by Western Blotting in E.Coli cell lysate.
Western Blotting Analysis: A 1:10,000 dilution of this antibody detected ICDH in 20 µL E.Coli cell lysate.
Western Blotting Analysis: A 1:10,000 dilution of this antibody detected ICDH in 20 µL E.Coli cell lysate.
Target description
~45 kDa observed. Uncharacterized bands may be observed in some lysate(s).
Physical form
Rabbit polyclonal antiserum with 0.05% sodium azide.
Unpurified
Storage and Stability
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Other Notes
Concentration: Please refer to lot specific datasheet.
Legal Information
GenBank is a registered trademark of United States Department of Health and Human Services
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Microbiology spectrum, 10(6), e0294922-e0294922 (2022-11-16)
The increasingly serious problem of bacterial drug resistance has led to the development of antivirulence agents. The Salmonella enterica serovar Typhimurium Salmonella pathogenicity island (SPI)-encoded type III secretion system (T3SS) and its effector proteins are important virulence factors for S.
Frontiers in microbiology, 12, 784926-784926 (2022-02-12)
Salmonella enterica serovar Typhimurium (S. Typhimurium) is a zoonotic pathogen that can cause food poisoning and diarrhea in both humans and animals worldwide. The Salmonella pathogenicity island (SPI) genes encoded type III secretion system (T3SS) is important for S. Typhimurium
Frontiers in cellular and infection microbiology, 12, 967149-967149 (2022-10-01)
New therapeutic strategies for clinical Salmonella enterica serovar Typhimurium (S. Typhimurium) infection are urgently needed due to the generation of antibiotic-resistant bacteria. Inhibition of bacterial virulence has been increasingly regarded as a potential and innovative strategy for the development of
Microbiology spectrum, 12(1), e0240623-e0240623 (2023-12-11)
Salmonella spp. remains a major worldwide health concern that causes significant morbidity and mortality in both humans and animals. The spread of antimicrobial resistant strains has declined the efficacy of conventional chemotherapy. Thus, novel anti-infection drugs or strategies are needed.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service