Skip to Content
Merck
All Photos(1)

Key Documents

8.14347

Sigma-Aldrich

Lithium tetrafluoroborate

for synthesis

Synonym(s):

Lithium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BF4Li
CAS Number:
Molecular Weight:
93.75
MDL number:
UNSPSC Code:
12352302
EC Index Number:
238-178-9
NACRES:
NA.22
Assay:
≥97.0% (acidimetric)
form:
powder

Quality Level

Assay

≥97.0% (acidimetric)

form

powder

potency

1638 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

mp

324 °C (decomposition)

storage temp.

2-30°C

SMILES string

[B]([Li])(F)(F)(F)F

InChI

1S/BF4.Li/c2-1(3,4)5;/q-1;+1

InChI key

UFXJWFBILHTTET-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 97.0 %
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service