Skip to Content
Merck
All Photos(1)

Key Documents

8.04512

Sigma-Aldrich

4-Hydroxy-4-methyl-2-pentanone

for synthesis

Synonym(s):

4-Hydroxy-4-methyl-2-pentanone, 2-Methyl-2-pentanol-4-one, Diacetone alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(OH)CH2COCH3
CAS Number:
Molecular Weight:
116.16
MDL number:
UNSPSC Code:
12352115
EC Index Number:
204-626-7
NACRES:
NA.22
form:
liquid
Assay:
≥98.0% (GC)

vapor pressure

1.1 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

640 °C

potency

2520 mg/kg LD50, oral (Rat)
13630 mg/kg LD50, skin (Rabbit)

expl. lim.

1.8-6.9 % (v/v)

bp

168 °C/1013 hPa

mp

-47 °C

transition temp

flash point 58 °C

density

0.937-0.941 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(CC(=O)C)(C)C

InChI

1S/C6H12O2/c1-5(7)4-6(2,3)8/h8H,4H2,1-3H3

InChI key

SWXVUIWOUIDPGS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.937 - 0.941
Identity (IR): passes test

Signal Word

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Repr. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service