Skip to Content
Merck

Skip To

673000

Vacuolin-1

≥95% (HPLC), solid, lysosome inhibitor, Calbiochem®

Synonym(s):

Vacuolin-1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C26H24IN7O
CAS Number:
Molecular Weight:
577.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥95% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Vacuolin-1, A cell-permeable triazine based compound that inhibits Ca2+-dependent fusion of lysosomes with the plasma membrane and the release of lysosomal content, without affecting enlargeosomes or other membrane-bound organelles.

Quality Segment

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

white

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Ic1cc(ccc1)\C=N\Nc2nc(nc(n2)N(c5ccccc5)c4ccccc4)N3CCOCC3

InChI

1S/C26H24IN7O/c27-21-9-7-8-20(18-21)19-28-32-24-29-25(33-14-16-35-17-15-33)31-26(30-24)34(22-10-3-1-4-11-22)23-12-5-2-6-13-23/h1-13,18-19H,14-17H2,(H,29,30,31,32)/b28-19+

InChI key

JMEJTSRAQUFNOP-TURZUDJPSA-N

General description

A cell-permeable triazine based compound that inhibits Ca2+-dependent fusion of lysosomes with the plasma membrane and the release of lysosomal content, without affecting enlargeosomes or other membrane-bound organelles. It does not disturb the organization of the actin or tubulin cytoskeletal network, nor does it interfere with membrane traffic along the constitutive pathway from the ER to the plasma membrane.
A cell-permeable triazine-based compound that inhibits Ca2+-dependent exocytosis of lysosomes induced by ionomycin or plasma membrane wounding. Does not affect the resealing process and the exocytosis of enlargeosomes. It also does not disturb the organization of the actin or tubulin cytoskeletal network or interfere with membrane traffic along the constitutive pathway from the ER to the plasma membrane.

Biochem/physiol Actions

Primary Target
Ca2+-dependent fusion of lysosomes with the plasma membrane
Product does not compete with ATP.
Reversible: no
Cell permeable: yes

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
373210365250171260
description

A cell-permeable triazine based compound that inhibits Ca2+-dependent fusion of lysosomes with the plasma membrane and the release of lysosomal content, without affecting enlargeosomes or other membrane-bound organelles.

description

The HDL Receptor SR-BI Inhibitor, BLT-1, also referenced under CAS 321673-30-7, controls the biological activity of HDL Receptor SR-B. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

description

Gö 6976, CAS 136194-77-9, is a cell-permeable, reversible, and ATP-competitive inhibitor of PKC (IC50 = 7.9 nM for rat brain). Exhibits selectively for PKCα (IC50 = 2.3 nM) and βI (IC50 = 6.2 nM).

description

AMPK Inhibitor, Compound C, CAS 866405-64-3, is a cell-permeable compound that inhibits KDR/VEGFR2, ALK2/BMPR-I, and AMPK kinase activities (IC50 = 25.1, 148, and 234.6 nM, respectively).

form

solid

form

solid

form

solid

form

solid

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage condition

protect from light, OK to freeze

storage condition

protect from light, OK to freeze

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters