Skip to Content
Merck
All Photos(1)

Documents

525276

Sigma-Aldrich

Phosphoramidon, Disodium Salt

Inhibits some metalloendopeptidases.

Synonym(s):

Phosphoramidon, Disodium Salt, N-(α-Rhamnopyranosyloxyhydroxyphosphinyl)-L-leucyl-L-tryptophan, 2Na

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32N3O10P · 2Na
Molecular Weight:
587.47
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 10 mg/mL
methanol: soluble
water: soluble

shipped in

ambient

storage temp.

2-8°C

General description

Effective concentration: 1-10 µM.
Inhibits some metalloendopeptidases. Highly specific inhibitor of thermolysin. Inhibits the conversion of big endothelin-1 to endothelin (IC50 = 4.6 µM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
thermolysin
Product does not compete with ATP.
Reversible: no
Target IC50: 4.6 µM against the conversion of big endothelin-1 to endothelin

Warning

Toxicity: Standard Handling (A)

Analysis Note

Single spot by TLC

Other Notes

Balwierczxak, J.L., et al. 1995. Biochem. Pharmacol. 49, 291.
Howell, S., et al. 1993. Biochem. J. 290, 159.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J L Balwierczak et al.
Biochemical pharmacology, 49(3), 291-296 (1995-01-31)
The enzyme responsible for the conversion of exogenous big endothelin-1 to endothelin-1 by porcine coronary arterial smooth muscle has been shown to be a metalloprotease. The potencies of eight metalloprotease inhibitors for this endothelin-converting enzyme were determined. CGS 25015, CGS
Jan Perner et al.
Frontiers in cellular and infection microbiology, 10, 563349-563349 (2020-12-15)
During feeding on vertebrate hosts, ticks secrete saliva composed of a rich cocktail of bioactive molecules modulating host immune responses. Although most of the proteinaceous fraction of tick saliva is of little immunogenicity, repeated feeding of ticks on mammalian hosts
S Howell et al.
The Biochemical journal, 290 ( Pt 1), 159-164 (1993-02-15)
Five membrane peptidase activities have been identified on cultured human osteoblast-like cells. These consisted of the four exopeptidases aminopeptidase-A, aminopeptidase-N, aminopeptidase-W and carboxypeptidase-M, and the endopeptidase, endopeptidase-24.11. The presence of endopeptidase-24.11 was confirmed immunochemically by immunofluorescent staining and by enzyme-linked

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service