Skip to Content
Merck
All Photos(1)

Key Documents

444213

Sigma-Aldrich

MMP-2, Proenzyme, Human Rheumatoid Synovial Fibroblast

Synonym(s):

Matrix Metalloproteinase 2, Gelatinase A, 72 kDa Type IV Collagenase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352202
NACRES:
NA.77

Assay

≥90% (SDS-PAGE)

Quality Level

form

liquid

specific activity

≥600 mU/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

solubility

aqueous buffer: soluble

foreign activity

other MMP activity, none detected

shipped in

wet ice

storage temp.

−70°C

General description

Native pro-MMP-2 from cultured, human rheumatoid synovial fibroblasts. Highly purified native enzyme derived from cell culture. Secreted by several cell types. Exists in a stable, but non-covalent 1:1 complex with TIMP-2. Requires activation prior to use.
Native pro-MMP-2 from cultured, human rheumatoid synovial fibroblasts. Secreted by several cell types. Requires activation just prior to use.
Note: 1 mU = 1 milliunit.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1.0 µmol 2,4-DNP-Pro-Gln-Gly-Ile-Ala-Gly-Gln-D-Arg-OH per min at 37°C, pH 7.0.

Physical form

In 200 mM NaCl, 50 mM Tris-HCl, 5 mM CaCl₂, 1 µM ZnCl₂, 0.05% Brij® 35 Detergent, 0.05% NaN₃, pH 7.0.

Preparation Note

Prepared from culture medium of human rheumatoid synovial fibroblasts that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Buisson-Leqendre, N., et al. 2004. Arth. Rheumat.50, 2151.
Morgunova, E., et al. 2002. Proc. Natl. Acad. Sci. USA99, 7414.
Kolkenbrock, H., et al. 1997. Biol. Chem. 378, 71.

Legal Information

Brij is a registered trademark of Croda International PLC
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service