1.25069
Fractogel® EMD TMAE (M)
MiniChrom Column Fractogel® TMAE (M), volume 5 mL, prepacked column
Synonym(s):
MiniChrom Column Fractogel® TMAE (M), 5 ml
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115711
Recommended Products
ligand
(Trimethylammoniumethyl)
material
polypropylene housing
product line
Fractogel®
form
prepacked column
parameter
≤8 bar pressure
~240 cm/hr max. flow rate
bed L
100 mm
bed volume
5 mL
column I.D.
8 mm
volume
5 mL
matrix active group
Methacrylate
mean particle size
40-90 μm
capacity
~100 mg, BSA binding capacity
application(s)
gene therapy
mAb
recombinant protein
separation technique
anion exchange
storage temp.
15-25°C
General description
8 x 100 mm
MiniChrom Pre-packed Columns with Fractogel® cation exchange resins are designed for the rapid evaluation of resins for bioprocess development. The results obtained for biomolecule separations are generally comparable to the results achieved using laboratory-scale columns (e.g. 100 x 16 mm ID).
In bioprocess design, the MiniChrom columns can be used to rapidly determine whether a target biomolecule is bound to and can be eluted from the Fractogel® resin, as well as determine whether an appropriate target yield may be achieved and whether major contaminants could potentially be removed.
MiniChrom columns are designed for process development, method optimization and parameter screening as well as sample preparation. MiniChrom columns are quality checked for HETP and asymmetry, providing superior reproducibility, excellent scalability, convenient comparison of different chromatography media.
MiniChrom columns are made from biocompatible polypropylene (PP) and polyethylene (PE) and are available in 1 and 5 mL bed volumes. The 8mm diameter columns allow for scale-up from 1 to 5mL column volumes, with a constant internal diameter. These columns are compatible with any chromatographic work station, HPLC system or peristaltic pump.
In bioprocess design, the MiniChrom columns can be used to rapidly determine whether a target biomolecule is bound to and can be eluted from the Fractogel® resin, as well as determine whether an appropriate target yield may be achieved and whether major contaminants could potentially be removed.
MiniChrom columns are designed for process development, method optimization and parameter screening as well as sample preparation. MiniChrom columns are quality checked for HETP and asymmetry, providing superior reproducibility, excellent scalability, convenient comparison of different chromatography media.
MiniChrom columns are made from biocompatible polypropylene (PP) and polyethylene (PE) and are available in 1 and 5 mL bed volumes. The 8mm diameter columns allow for scale-up from 1 to 5mL column volumes, with a constant internal diameter. These columns are compatible with any chromatographic work station, HPLC system or peristaltic pump.
Features and Benefits
MiniChrom Column Fractogel® TMAE (M), 5 ml enables:
- High protein loading conditions resulting in good selectivity and resolution with the tentacle technology
- Excellent binding to large viruses at increased loading
- Homogenous binding with high selectivity and purity
- Reduce operating costs and resin replacement with enduring durability - allowing for multiple cycles of column regeneration and sanitization
- Compatibility with 2.5 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl storage solution
Legal Information
FRACTOGEL is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service