Skip to Content

119136

Adenosine A1 Receptor Agonist II, CCPA

Synonym(s):

Adenosine A1 Receptor Agonist II, CCPA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H20ClN5O4
CAS Number:
Molecular Weight:
369.80
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥99% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99% (HPLC)

Quality Segment

form

solid

potency

2.3 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

white

solubility

DMSO: 100 mM, water: 5 mM

shipped in

ambient

storage temp.

2-8°C

SMILES string

Clc1nc2[n](cnc2c(n1)NC4CCCC4)[C@@H]3O[C@@H]([C@H]([C@H]3O)O)CO

InChI

1S/C15H20ClN5O4/c16-15-19-12(18-7-3-1-2-4-7)9-13(20-15)21(6-17-9)14-11(24)10(23)8(5-22)25-14/h6-8,10-11,14,22-24H,1-5H2,(H,18,19,20)/t8-,10-,11-,14-/m1/s1

InChI key

XSMYYYQVWPZWIZ-IDTAVKCVSA-N

General description

An adenosine analog that acts as a selective, high affinity agonist for Adenosine A1 receptor (A1R) (Ki = 800 pM; 2.3 µM, 18.8 µM, and 42 nM for human A1, A2A, A2B, A3 respectively). Shown to be centrally active following systemic administration and exhibits anti-convulsive effects. Also reported to inhibit adenylate cyclase activity in rat fat cell membrane (IC50 = 33 nM). Diminishes respiratory rhythm, reduces heart rate, and enhances the activity of KATP channels. Reduces norepinephrine release by about 50% in untreated and pertussis toxin treated hearts.

Biochem/physiol Actions

Primary Target
A1
Reversible: yes

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Burgdorf, C. et al. 2005. J Cardiovasc Pharmacol.45, 1.
Klotz, K. N., et al. 2000. Naunyn Schmiedebergs Arch Pharmacol. 362, 382.
Mironov, S., et al. 1990. J Neurophysiol.81, 1.
Monopoli, A., et al. 1994.Arzneimittelforschung.44, 1305.
Concas, A., et al. 1993. J Pharmacol Exp Ther.267, 844.
Lohse, M. J., et al. 1988. Nauryn Schmiedebergs Arch Pharmacol.337, 687.
Coffin, V. L., et al. 1987. J Pharmacol Exp Ther.241, 76.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library