1.03672
Cholesterol from wool fat
extra pure, powdered
Pharma Manufacturing
Synonym(s):
Cholesterol from wool fat
About This Item
Recommended Products
Quality Level
form
powder
bp
200 °C (Decomposition)
mp
141 °C
solubility
insoluble >0.001 g/L
density
1.05 g/cm3 at 20 °C
storage temp.
15-25°C
SMILES string
O[C@H]1CC[C@@]2([C@@H]3[C@H]([C@H]4[C@@]([C@H](CC4)[C@@H](CCCC(C)C)C)(CC3)C)CC=C2C1)C
InChI
1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1
InChI key
HVYWMOMLDIMFJA-DPAQBDIFSA-N
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service