Skip to Content
Merck

1.01885

SAFC

Cellvento® CHO-200

Chemically defined cell culture medium

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

Quality Level

description

shipped with cooling packs

product line

Cellvento®

form

dry powder

technique(s)

cell culture | mammalian: suitable (suspension)

storage temp.

2-8°C

General description

Cellvento® CHO-200 chemically defined cell culture medium has been specially developed for the growth of Chinese Hamster Ovary (CHO) cells and the expression of monoclonal antibodies and recombinant proteins in suspension culture. The formulation is of non-animal origin, chemically defined and contains no hydrolysates or components of unknown composition. Cellvento® CHO-200 medium has been formulated without L-glutamine, hypoxanthine, and thymidine to keep flexibility in applications. It is available in dry powder form or as ready-to-use medium to fit to different experimental set-ups.

Features and Benefits

Cellvento® CHO-200 medium and its feeds have been designed for use in the CHO-S GS expression system in cell suspension culture, but may be suitable for other CHO-S cell lines.
  • Cellvento® CHO-200 medium should be used for cell adaptation and cell bank generation.
  • Cellvento® CHO-200 medium is suitable for use in seed train expansion.
  • Cellvento® products allow for flexibility in feed and feed supplement optimization of fed-batch processes.
This product is intended for research or further manufacturing but not for human or therapeutic use.

Quantity

Formulated to contain 115.8 grams of powder per liter of medium.

Legal Information

CELLVENTO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page summarizes key considerations related to regulatory expectations for biomanufacturing including chemicals and polymeric materials. For chemicals, topics include nitrosamines and elemental impurities, and for polymeric materials such as filters and single-use assemblies, the focus is qualification and extractable and leachable testing.

This page summarizes key considerations related to regulatory expectations for biomanufacturing including chemicals and polymeric materials. For chemicals, topics include nitrosamines and elemental impurities, and for polymeric materials such as filters and single-use assemblies, the focus is qualification and extractable and leachable testing.

This page summarizes key considerations related to regulatory expectations for biomanufacturing including chemicals and polymeric materials. For chemicals, topics include nitrosamines and elemental impurities, and for polymeric materials such as filters and single-use assemblies, the focus is qualification and extractable and leachable testing.

This page summarizes key considerations related to regulatory expectations for biomanufacturing including chemicals and polymeric materials. For chemicals, topics include nitrosamines and elemental impurities, and for polymeric materials such as filters and single-use assemblies, the focus is qualification and extractable and leachable testing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service