Skip to Content
Merck
All Photos(3)

Documents

07-1406

Sigma-Aldrich

Anti-GLUT-5 Antibody, a.a. 490-502

serum, from rabbit

Synonym(s):

Fructose transporter, Glucose transporter type 5, small intestine, glucose transporter-like protein 5, solute carrier family 2 (facilitated glucose transporter),

member 5, solute carrier family 2 (facilitated glucose/fructose

transporte

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, guinea pig, mouse

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Slc2A5(56485)

Related Categories

General description

A cytochalasin B-sensitive carrier located in the membrane, GLUT-5 appears to be primarily functional as a fructose transporter. It is expressed in the small intestine, and is also found in the skeletal muscle, kidney and adipose tissue but at significantly reduced levels.

Specificity

Recognizes GLUT-5.

Immunogen

Epitope: C-terminus
Synthetic peptide corresponding to the C-terminus of rat GLUT-5 (aa 490-502) coupled to KLH. The immunogen sequence used is only 53% conserved in mouse.

Application

Anti-GLUT-5 Antibody, a.a. 490-502 detects level of GLUT-5 & has been published & validated for use in WB.
Research Category
Signaling
Research Sub Category
Insulin/Energy Signaling

Quality

Evaluated by Western Blot in Mouse kidney lysate.
Western Blot Analysis: A 1:1000 dilution of this antibody detected GLUT-5 in 10 µg of mouse kidney lysates.

Target description

Approx. 54 kDa

Linkage

Replaces: AB1348

Physical form

Liquid containing 0.05% sodium azide.
Unpurified rabbit serum

Storage and Stability

Store at -20°C in undiluted aliquots for up to 1 year from date of receipt. Avoid repeated freeze-thaw cycles.

Analysis Note

Control
Mouse kidney lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irene Zwarts et al.
Scientific reports, 9(1), 9299-9299 (2019-06-28)
Fructose has become a major constituent of our modern diet and is implicated as an underlying cause in the development of metabolic diseases. The fructose transporter GLUT5 (SLC2A5) is required for intestinal fructose absorption. GLUT5 expression is induced in the
High Fructose Corn Syrup Accelerates Kidney Disease and Mortality in Obese Mice with Metabolic Syndrome.
Andres-Hernando, et al.
Biomolecules, 13 (2023)
Ana Andres-Hernando et al.
Cell metabolism, 32(1), 117-127 (2020-06-06)
Intake of fructose-containing sugars is strongly associated with metabolic syndrome. Compared with other sugars, dietary fructose is uniquely metabolized by fructokinase. However, the tissue-specific role of fructokinase in sugar-induced metabolic syndrome, and the specific roles of glucose and fructose in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service