Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
SK703, monoclonal
Species reactivity:
mouse, human
Application:
WB
Citations:
5
biological source
rabbit
Quality Level
antibody form
culture supernatant
antibody product type
primary antibodies
clone
SK703, monoclonal
species reactivity
mouse, human
manufacturer/tradename
Upstate®
technique(s)
western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
phosphorylation (pSer473)
Gene Information
human ... AKT1(207)
General description
60kDa
The serine/threonine kinase Akt family contains several members, including Akt1 (also designated PKBα or RacPK), Akt2 (also designated PKB-β or RacPK-β) and Akt3 (also designated PKB-γ or thyoma viral proto-oncogene 3). Akt1 and Akt2 are activated by PDGF stimulation. This activation is dependent on PDGFR-β tyrosine residues 740 and 751, which bind the 85 kDa subunit of the phosphatidylinositol 3-kinase (PI 3-kinase) complex. Activation of Akt1 by insulin or insulin-growth factor-1 (IGF-1) results in phosphorylation of both Thr 308 and Ser 473. Phosphorylation of both residues is important to generate a high level of Akt1 activity, and the phosphorylation of Thr 308 is not dependent on phosphorylation of Ser 473 in vivo. Thus, Akt proteins become phosphorylated and activated in insulin/IGF-1- stimulated cells by an upstream kinase(s). The activation of Akt1 and Akt2 is inhibited by the PI kinase inhibitor wortmannin. Taken together, this data strongly suggests that the protein signals downstream of the PI kinases.
Immunogen
Epitope: Phosphorylated Ser473
Peptide containing a pSer that corresponds to amino acid position 473 of human Akt1/PKBa
Application
Anti-Akt1 Antibody, clone SK703 detects level of phospho-Akt1/PKBα (Ser473) & has been published & validated for use in WB.
Research Category
Signaling
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling
PI3K, Akt, & mTOR Signaling
Biochem/physiol Actions
Predicted cross-reactivity with rat, bovine, chicken, and Xenopus based on immunogen sequence homology.
Recognizes phosphorylated Akt1/PKBα, Mr 60 kDa. Cross-reactivity to phosphorylated Akt2 and Akt3 likely based on sequence homology.
Physical form
Cultured supernantant in 0.05% sodium azide
Preparation Note
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Analysis Note
Control
Untreated and PDGF treated NIH-3T3 fibroblasts.
Untreated and PDGF treated NIH-3T3 fibroblasts.
routinely evaluated by immunoblot on lysates from mouse NIH-3T3 fibroblasts treated with PDGF
Other Notes
Replaces: 05-736
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Explore all of our products under Anti-phospho-Akt1/PKBα (Ser473) Antibody, clone SK703, rabbit monoclonal
— or —
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Meiwan Cao et al.
Journal of immunology (Baltimore, Md. : 1950), 198(5), 1887-1899 (2017-01-29)
In myelodysplastic syndromes (MDS), functional defects of neutrophils result in high mortality because of infections; however, the molecular basis remains unclear. We recently found that miR-34a and miR-155 were significantly increased in MDS neutrophils. To clarify the effects of the
Yung-Hung Hsieh et al.
Nutrients, 10(9) (2018-09-12)
The anti-inflammatory effect of ginsenoside Rh2 (GRh2) has labeled it as one of the most important ginsenosides. The purpose of this study was to identify the anti-inflammatory and antioxidant effects of GRh2 using a lipopolysaccharide (LPS) challenge lung-injury animal model.
Short high-fat diet interferes with the physiological maturation of the late adolescent mouse heart.
Heidi Hynynen et al.
Physiological reports, 8(13), e14474-e14474 (2020-07-10)
Dietary fats are essential for cardiac function. The metabolites of fats known as fatty acids provide most of the energy for cardiac tissue, serve as building blocks for membranes and regulate important signaling cascades. Despite their importance, excess fat intake
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 04-736 | 04053252372315 |