Skip to Content
Merck
All Photos(1)

Key Documents

880220P

Avanti

18:0 PEG5000 PE

Avanti Research - A Croda Brand 880220P, powder

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-5000] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 g (880220P-1g)
pkg of 1 × 200 mg (880220P-200mg)
pkg of 1 × 25 mg (880220P-25mg)
pkg of 1 × 500 mg (880220P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880220P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC([(OCH2CH2)113OCH3])=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[NH4+]

General description

18:0 PEG5000 PE is a polyethylene glycol (PEG) conjugated phospholipid. PEGylated phospholipids are amphiphilic polymers.

Application

18:0 PEG5000 PE is suitable for particle formulation used in gene delivery study.

Biochem/physiol Actions

18:0 PEG5000 PE is useful in liposome formulation and used as drug carriers.

Packaging

20 mL Clear Glass Screw Cap Vial (880220P-1g)
20 mL Clear Glass Screw Cap Vial (880220P-500mg)
5 mL Amber Glass Screw Cap Vial (880220P-200mg)
5 mL Amber Glass Screw Cap Vial (880220P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Degradable terpolymers with alkyl side chains demonstrate enhanced gene delivery potency and nanoparticle stability
Eltoukhy AA, et al.
Advanced Materials, 25(10), 1487-1493 (2013)
The
Micro- and Nanoengineering of the Cell Surface, 110-110 (2014)
Oblique Self-Assemblies and Order- Order Transitions in Polypeptide Complexes with PEGylated Triple-Tail Lipids
Hanski S, et al.
Biomacromolecules, 11(12), 3440-3447 (2010)
Cell culture studies of a carborane cholesteryl ester with conventional and PEG liposomes
Peacock GF, et al.
Drug delivery, 10(1), 29-34 (2003)
Richard J Browning et al.
Langmuir : the ACS journal of surfaces and colloids, 36(2), 609-617 (2019-12-20)
Microbubbles stabilized by an outer lipid shell have been studied extensively for both diagnostic and therapeutic applications. The shell composition can significantly influence microbubble behavior, but performing quantitative measurements of shell properties is challenging. The aim of this study is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service