Skip to Content
Merck
All Photos(1)

Key Documents

800818C

Avanti

18:0-20:4 DG

1-stearoyl-2-arachidonoyl-sn-glycerol, chloroform

Synonym(s):

1-octadecanoyl-2-(9Z,12Z-octadecadienoyl)-sn-glycerol; DG(18:0/18:2(9Z,12Z)/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H72O5
CAS Number:
Molecular Weight:
645.01
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 5 mL (800818C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 800818C

concentration

2 mg/mL (800818C-10mg)

lipid type

neutral lipids
neutral glycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

O([C@H](COC(=O)CCCCCCCCCCCCCCCCC)CO)C(=O)CCC\C=C/C\C=C/C\C=C/C\C=C/CCCCC

InChI

1S/C41H72O5/c1-3-5-7-9-11-13-15-17-19-20-22-24-26-28-30-32-34-36-41(44)46-39(37-42)38-45-40(43)35-33-31-29-27-25-23-21-18-16-14-12-10-8-6-4-2/h11,13,17,19,22,24,28,30,39,42H,3-10,12,14-16,18,20-21,23,25-27,29,31-38H2,1-2H3/b13-11-,19-17-,24-22-,30-28-/t39-/m0/s1

InChI key

NSXLMTYRMFVYNT-IUJDHQGTSA-N

General description

18:0-20:4 DG or 1-stearoyl-2-arachidonoyl-sn-glycerol is a diglyceride. Diglycerides contain two fatty acids linked to the glycerol backbone with an ester bond.

Application

18:0-20:4 DG or 1-stearoyl-2-arachidonoyl-sn-glycerol has been used to study its effect on conventional protein kinase C (cPKC) and novel protein kinase C (nPKC) isozymes in vitro. It has also been used as a substrate for the measurement of diacylglycerol kinase δ (DGK#948;) and DGKη1 activity in vitro.

Biochem/physiol Actions

In biochemical signaling, diacylglycerol (DAG) functions as a second messenger signaling lipid, and is a product of the hydrolysis of the phospholipid PIP2 (phosphatidylinositolbisphosphate) by the enzyme phospholipase C (PLC) (a membrane-bound enzyme) that, through the same reaction, produces inositol trisphosphate (IP3). Although inositol trisphosphate (IP3) diffuses into the cytosol, DAG remains within the plasma membrane due to its hydrophobic properties. IP3 stimulates the release of calcium ions from the smooth endoplasmic reticulum, whereas DAG is a physiological activator of protein kinase C (PKC). The production of DAG in the membrane facilitates translocation of PKC from the cytosol to the plasma membrane.
Diacylglycerol mimicks the effects of the tumor-promoting compounds phorbol esters.

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (800818C-10mg)

Storage and Stability

Diacylglycerols are conveniently stored in chloroform solutions in glass vials with PTFE-lined caps at -20°C. Under these conditions acyl migration is minimal. Avoid plastic when handling chloroform solutions.

Other Notes

Delivery to cells:
Dry samples of diacylglycerol in chloroform, using a stream of nitrogen. Dissolve the residue in an appropriate volume of ethanol or DMSO, then dilute to the desired aqueous medium.
Effective concentration:
Most biological responses saturate at 20 to 250 μM sn-1,2-dioctanoylglycerol. Only sn-1,2 isomers appear to be active.
Precaution: Since short chain Diacylglycerols mimic effects of the tumor-promoting phorbol diesters in a number of biological systems, extra care should be employed in their handling. Treatment of solutions, vessels and other articles with 1N NaOH before washing or discarding will destroy diacylglycerols.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert C Murphy et al.
Journal of lipid research, 60(2), 219-226 (2019-01-05)
Leukotrienes (LTs) are autacoids derived from the precursor arachidonic acid (AA) via the action of five-lipoxygenase (5-LO). When inflammatory cells are activated, 5-LO translocates to the nuclear membrane to initiate oxygenation of AA released by cytosolic phospholipase A2 (cPLA2) into
Hiromichi Sakai et al.
The Journal of biological chemistry, 289(38), 26607-26617 (2014-08-13)
Decreased expression of diacylglycerol (DG) kinase (DGK) δ in skeletal muscles is closely related to the pathogenesis of type 2 diabetes. To identify DG species that are phosphorylated by DGKδ in response to high glucose stimulation, we investigated high glucose-dependent
Yuuna Kamiya et al.
Biochemistry and biophysics reports, 7, 361-366 (2016-07-20)
A variety of diacylglycerol (DG) molecular species are produced in stimulated cells. Conventional (α, βII and γ) and novel (δ, ε, η and θ) protein kinase C (PKC) isoforms are known to be activated by DG. However, a comprehensive analysis
Sruthi S Balakrishnan et al.
Journal of cell science, 131(15) (2018-07-08)
The activation of phospholipase C (PLC) is a conserved mechanism of receptor-activated cell signaling at the plasma membrane. PLC hydrolyzes the minor membrane lipid phosphatidylinositol 4,5-bisphosphate [PI(4,5)P2], and continued signaling requires the resynthesis and availability of PI(4,5)P2 at the plasma
Maria Saliakoura et al.
Oncogene, 39(14), 2948-2960 (2020-02-09)
Enhanced prostaglandin production promotes the development and progression of cancer. Prostaglandins are generated from arachidonic acid (AA) by the action of cyclooxygenase (COX) isoenzymes. However, how cancer cells are able to maintain an elevated supply of AA for prostaglandin production

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service