Skip to Content
Merck
All Photos(1)

Key Documents

791649C

Avanti

18:1-d9 SM

Avanti Research - A Croda Brand

Synonym(s):

N-oleoyl(d9)-D-erythro-sphingosylphosphorylcholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H72 D9N2O6P
CAS Number:
Molecular Weight:
738.12
MDL number:
UNSPSC Code:
41141825
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (791649C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL (791649C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

18:1-d9 SM (N-oleoyl(d9)-D-erythro-sphingosylphosphorylcholine) is a deuterated derivative of 18:1-SM (N-oleoyl-D-erythro-sphingosylphosphorylcholine). 18:1- SM is a sphingomyelin which is present in aqueous humor. 18:1-SM or oleoyl-sphingomyelin, comprises quaternary ammonium group. 18:1-SM or oleoyl-sphingomyelin due to its miscible property is incapable of sequestering with cholesterol-rich domains.
This deuterium labeled lipid is also available in a formulation of multiple lipid standards. The SPLASH LIPIDOMIX Standard includes all of the major lipid classes at ratios relative to human plasma, allowing one easy internal standard solution to be added to every sample.

Application

18:1-d9 SM (N-oleoyl(d9)-D-erythro-sphingosylphosphorylcholine) has been used as an extraction buffer component for lipid isolation from raft and non-raft fractions of synaptic vesicles (SV) from rat brain. It may be used as an internal standard to spike serum lipid extracts for liquid chromatography-mass spectrometry (LC/MS) analysis and ultra-high-performance supercritical fluid chromatography with quadrupole-time-of-flight mass spectrometry (UHPSFC/QTOF-MS) for signal normalization and quantification of pulmonary artery lipids form porcine.

Packaging

5 mL Clear Glass Sealed Ampule (791649C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
LIPIDOMIX is a trademark of Avanti Polar Lipids, LLC
SPLASH is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ayman J Aljohani et al.
Molecular vision, 19, 1966-1984 (2013-09-27)
To determine the differential profiles of sphingomyelin, sphingoid base, sphingoid base-1-phosphate and ceramide lipid species and their quantitative differences between control and glaucomatous aqueous humor (AQH) derived from human donors. AQH from control and primary open-angle glaucoma donors was collected
Richard M Epand et al.
Chemistry and physics of lipids, 132(1), 37-46 (2004-11-09)
Sphingomyelin from biological membranes forms segregated domains with cholesterol in fluid bilayers. However, a synthetic form of sphingomyelin with an oleoyl chain linked to sphingosine is not incorporated into cholesterol-rich domains. We have studied the properties of mixtures of oleoyl-sphingomyelin
Alicia Gil-Ramirez et al.
Journal of chromatography. A, 1592, 173-182 (2019-02-03)
Pulmonary artery grafts are needed as cardiovascular bioprosthetics. For successful tissue recellularization after transplantation, lipids have to be removed from the donor artery. Developing a selective process to remove lipids without damaging the extracellular matrix greatly depends on knowing the
John Williams et al.
Bioanalysis, 12(3), 143-158 (2020-02-14)
Aim: Very long chain fatty acids (VLCFAs) have been identified as biomarkers for several peroxisomal disorders necessitating the need for reliable biomarker assays in particular C20, C22, C24, C26 in cerebrospinal fluid (CSF). Until now no absolute quantitation assay for
Michael S Gardner et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1118-1119, 137-147 (2019-04-30)
Progress toward better diagnosis and treatment of lipid metabolism-related diseases requires high throughput approaches for multiplexed quantitative analysis of structurally diverse lipids, including phospholipids (PLs). This work demonstrates a simplified "one-pot" phospholipid extraction protocol, as an alternative to conventional liquid-liquid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service