Skip to Content
Merck
All Photos(1)

Key Documents

Z743910

ART® SoftFit L Reload System

tip design ART 20L, size 20 μL, racked, sterile

Synonym(s):

aerosol barrier tips, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip, sterile barrier pipette tip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02

sterility

sterile

feature

racked
tip design ART 20L

packaging

pkg of 5X960 ea

manufacturer/tradename

Molecular BioProducts 2749

technique(s)

PCR: suitable

size

20 μL

Looking for similar products? Visit Product Comparison Guide

General description

“SoftFit-L”pipette tips enable you to pipette samples with the accuracy and precision you expect from your pipette, every time.

Features and Benefits

  • Available in low retention variants.
  • Sterile tips are certified to be RNase, DNase, and pyrogen free.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service