Skip to Content
Merck
All Photos(1)

Key Documents

Z743815

ART® Non-Filtered Low Retention Tip

HLT uLtra micro 20uL, size 20 μL, racked, sterile

Synonym(s):

aerosol barrier tips, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip, sterile barrier pipette tip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121603
NACRES:
NB.02

Pricing and availability is not currently available.

sterility

sterile

feature

HLT uLtra micro 20uL
racked

packaging

pkg of 5X960 ea

manufacturer/tradename

Molecular BioProducts 3521-05

technique(s)

PCR: suitable

size

20 μL

Looking for similar products? Visit Product Comparison Guide

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service