Z530360
Aldrich® in-line oil bubbler
joint: ST/NS 19/22
Synonym(s):
bubblers, in-line bubblers, scientific bubblers
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
Joint
joint: ST/NS19/22
capacity
10 mL
Related Categories
General description
For precise N2 pressure control during inert atmosphere reactions. Connect reaction vessel to in-line joint or use with a ballast bulb to keep pressure constant.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)