Key Documents
SYNF341200
Synthware two neck pear shaped flask
200 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
Synonym(s):
Synthware F341200, 2-neck pear shape flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
material
borosilicate glass flask
pear-shaped
two-neck
Joint
center joint: ST/NS14/20
side joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F341200
capacity
200 mL
200 mL
General description
Pear shape with joints. Heavy wall design.
Legal Information
Synthware is a trademark of Kemtech America Inc.