Merck
SYNC214550
Synthware Friedrichs condenser
top joint: ST/NS 24/40, bottom joint: ST/NS 45/50
Synonym(s):
Synthware C214550, glass Friedrichs condenser
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.51
UNSPSC Code:
41121800
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
joint
bottom joint: ST/NS45/50, joint: ST/NS top24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C214550
General description
With a joint at 15º angle.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Still not finding the right product?
Explore all of our products under Synthware Friedrichs condenser
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Global Trade Item Number
SKUGTIN
SYNC214550-1EA04061832190846