Skip to Content
Merck
All Photos(2)

Key Documents

GF62158899

Indium

wire reel, 1m, diameter 0.25mm, as drawn, 99.999%

Synonym(s):

Indium, IN005110

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
In
CAS Number:
Molecular Weight:
114.82
MDL number:
UNSPSC Code:
12141719
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 25 °C)

Assay

99.999%

form

wire

manufacturer/tradename

Goodfellow 621-588-99

resistivity

8.37 μΩ-cm

L × diam.

1 m × 0.25 mm

mp

156.6 °C (lit.)

density

7.3 g/mL at 25 °C (lit.)

SMILES string

[In]

InChI

1S/In

InChI key

APFVFJFRJDLVQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Effect of indium and its compounds on the body (review of the literature)].
Iu I Pokhodzeĭ
Gigiena truda i professional'nye zabolevaniia, (4)(4), 43-45 (1984-04-01)
Kazuyuki Omae et al.
International archives of occupational and environmental health, 84(5), 471-477 (2010-10-05)
The present review is aimed to introduce an new occupational lung disease, Indium Lung. We searched case reports and epidemiological studies concerning indium-related lung diseases and reviewed. Up to March, 2010, 7 cases of interstitial pneumonia in Japanese indium-exposed workers
A M Peters et al.
Seminars in thrombosis and hemostasis, 9(2), 100-114 (1983-04-01)
The application of 111In-oxine to platelet labeling has contributed to the understanding of platelet kinetics along three lines: 1. It allows the measurement of new parameters of splenic function, such as the intrasplenic platelet transit time, which has shed new
The significance of chromosomal aberrations in indium-111-labeled lymphocytes.
M L Thakur et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 25(8), 922-927 (1984-08-01)
Indium-111 imaging in osteomyelitis and neuroarthropathy. Review and case report.
L M Techner et al.
Journal of the American Podiatric Medical Association, 76(1), 23-29 (1986-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service