DWK209666
Wheaton® Leak Resistant Bottle
capacity 60 mL, polypropylene bottle, natural bottle, wide-mouth bottle, bottle diam. × H 39 mm × 88 mm, 28-410
Synonym(s):
209666
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Recommended Products
material
natural bottle
polypropylene bottle
wide-mouth bottle
packaging
pkg of 72 ea
manufacturer/tradename
WHEATON® 209666
bottle diam. × H
39 mm × 88 mm
capacity
60 mL
application(s)
PFAS testing
Looking for similar products? Visit Product Comparison Guide
Related Categories
Features and Benefits
Superior double seal closure for leak resistance
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service