Skip to Content
Merck
All Photos(1)

Key Documents

C58800

Sigma-Aldrich

5-Chloro-2-nitrobenzaldehyde

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)CHO
CAS Number:
Molecular Weight:
185.56
Beilstein:
1910197
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

form

powder

impurities

<15% 3-chloro-4-nitrobenzaldehyde
<3% sulfuric acid
<5% 5-chloro-2-nitrobenzoic acid

mp

65-69 °C (lit.)

SMILES string

[H]C(=O)c1cc(Cl)ccc1[N+]([O-])=O

InChI

1S/C7H4ClNO3/c8-6-1-2-7(9(11)12)5(3-6)4-10/h1-4H

InChI key

SWGPIDCNYAYXMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zabiulla et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 95, 419-428 (2017-09-02)
Disrupted redox balance is implicated in multiple pathologies including malignant progression and tumor angiogenesis. In this investigation, we report the design and development of novel and effective ROS detoxifying azo-hydrazone molecules targeting malignant pathologies and neoangiogenesis. A series of azo-derivatives

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service