Skip to Content
Merck
All Photos(1)

Key Documents

BR781720

BRAND® microplate BRANDplates®, immunoGrade

size 96 wells, size 330 μL, round bottom, transparent

Synonym(s):

BRANDplates®, Immunology, Microplates, high binding, immunoGrade

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
colorless wells
polystyrene
round bottom
size:
330 μL
96 wells

material

colorless wells
polystyrene
round bottom

sterility

non-sterile

feature

lid: no
skirt (U-bottom)

packaging

pack of 100 ea

manufacturer/tradename

BRAND 781720

size

330 μL
96 wells

working volume

330 μL

color

transparent

suitability

suitable for (bacteriological assays)

binding type

high binding surface

Looking for similar products? Visit Product Comparison Guide

General description

The BRAND® Microplate BRANDplates® ImmunoGrade is specifically designed for high-binding capacity, optimized for the immobilization of IgG. This microplate offers superior binding for molecules with mixed hydrophilic and hydrophobic regions, making it the surface of choice for the majority of standard ELISAs and solid-phase immunoassays. Comparable to high-binding plates from other manufacturers, it is made from durable polystyrene (PS) and is non-sterile. Additionally, it is BIO-CERT® CERTIFIED QUALITY, ensuring it is free from human DNA, RNase, DNase, and pyrogens, making it ideal for sensitive applications in research and diagnostics. This microplate is an essential tool for achieving reliable and reproducible results in immunological assays.

Features and Benefits

  • Optimal surfaces for optimal results
  • Reliable quality from the cleanroom
  • Tested by independent laboratories

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service