BR474410
BRAND® plastic staining trough, Schiefferdecker pattern
with lid
Synonym(s):
Microscopy, Staining, Staining Accessories
All Photos(1)
About This Item
UNSPSC Code:
41122800
NACRES:
MA.01
Recommended Products
material
PMP
feature
with lid
manufacturer/tradename
BRAND 474410
L × W
76 mm × 26 mm , holds 20 slides
Related Categories
General description
The BRAND® Plastic Staining Trough in the Schiefferdecker pattern is designed for efficient staining of up to 20 slides, organized in 10 pairs back-to-back. With dimensions of 76 x 26 mm, this trough provides ample space for optimal staining while allowing easy access to each slide. Its robust construction ensures durability and reliability in laboratory settings, making it an essential tool for histology and microscopy applications.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)