BAF367750050
SP Bel-Art® Sterileware™ Upright Handle Dippers
blue plastic, capacity 50 mL (1¾ oz), sterile
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
Recommended Products
material
blue plastic
sterility
sterile
packaging
pack of 40 ea
manufacturer/tradename
Bel-Art F36775-0050
L
10.25 in. (260 mm)
capacity
50 mL (1¾ oz)
cup diam.
1.875 in. (48 mm)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Available in blue or white to allow for effective sample contrast, these sterile ladles are useful for sampling liquids and creams. Upright handle allows for sampling from above container, curved handle end provides secure withdrawal of sample . Fits into 2” openings such as a 45-gallon drum bunghole.
Features and Benefits
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- White high density polyethylene (HDPE)
- Blue polypropylene (PP)
- Heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10^-6)
- Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service