Skip to Content
Merck
All Photos(3)

Documents

936170

Sigma-Aldrich

EPhos Pd G3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H68NO4PPdS
CAS Number:
Molecular Weight:
904.53
UNSPSC Code:
12352142
NACRES:
NA.21

form

powder

Quality Level

reaction suitability

reaction type: Cross Couplings

color

gray

functional group

(Palladium)
phosphine

SMILES string

O=S(=O)([O-][Pd+2]1([C-]=2C=CC=CC2C=3C=CC=CC3[NH2]1)[P](C=4C(OC(C)C)=CC=CC4C=5C(=CC(=CC5C(C)C)C(C)C)C(C)C)(C6CCCCC6)C7CCCCC7)C

General description

EPhos Pd G3 is a third generation (G3) Buchwald palladacycle precatalyst with EPhos ligand for Pd-catalyzed C–N cross-coupling between primary amines and aryl halides to form 2-arylaminooxazoles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service