912905
Enantioprobe (R)-1
≥95%
Synonym(s):
(R)-3-(3-(But-3-yn-1-yl)-3H-diazirin-3-yl)-1-(2-phenylpyrrolidin-1-yl)propan-1-one, Fully functionalized enantioprobe, Ligandability probe
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C18H21N3O
Molecular Weight:
295.38
UNSPSC Code:
12161600
Assay
≥95%
form
liquid
storage temp.
−20°C
Application
Enantioprobe (R)-1 is one of 16 fully functionalized, enantiomeric fragment probes developed in the Cravatt lab. Combining fragment-based ligand discovery (FBLD) with chemical proteomics, the enantioprobe library assesses ligandability across proteomes. Each enantioprobe contains a structurally variable fragment for interaction with proteins, photoactivatable diazirine for UV-induced covalent protein labeling, and bioorthogonal alkyne handle for detection, enrichment, and identification. Of the eight enantiomeric pairs, each differs by one stereocenter, and comparing stereoselective fragment-protein interactions between the pairs simplifies validation of authentic protein-binding events. Enantioprobe (R)-1′s paired fragment is available as Enantioprobe (S)-1 (cat# 912417).
Together, the 16 enantioprobes support ligandability studies in living cells, a significant method for development of chemical probes and lead discovery efforts to find binders for traditionally ″undruggable″ protein targets. This strategy is also compatible with multiplexing for higher throughput.
Together, the 16 enantioprobes support ligandability studies in living cells, a significant method for development of chemical probes and lead discovery efforts to find binders for traditionally ″undruggable″ protein targets. This strategy is also compatible with multiplexing for higher throughput.
Supporting reagents:
- Fluorophore-conjugated azide tags for SDS-PAGE in-gel profiling: 910147, 760757, 760765, 777315, 777323
- Azide-biotin tags for enrichment: 901765, 914134, 914460, 762024, 900912, 900891, QBD10825)
- Azide capture solid supports: 900957, 742627
- Streptavidin for biotin enrichment: S6940, E5529, E2513, 69203, 16-126, S1638
- Mass spectrometry and Multiplexing
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yujia Wang et al.
Nature chemistry, 11(12), 1113-1123 (2019-10-30)
A fundamental challenge in chemical biology and medicine is to understand and expand the fraction of the human proteome that can be targeted by small molecules. We recently described a strategy that integrates fragment-based ligand discovery with chemical proteomics to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service