724017
1H-Indazole-5-boronic acid
≥95%
Synonym(s):
1H-Indazol-5-yl-5-boronic acid, Indazole-5-boronic acid
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C7H7BN2O2
CAS Number:
Molecular Weight:
161.95
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥95%
form
solid
mp
175-180 °C
storage temp.
2-8°C
SMILES string
OB(O)c1ccc2[nH]ncc2c1
InChI
1S/C7H7BN2O2/c11-8(12)6-1-2-7-5(3-6)4-9-10-7/h1-4,11-12H,(H,9,10)
InChI key
CLVPGJWAMIADSY-UHFFFAOYSA-N
Application
Reactant for:
- Preparation of thieno[3,2-d]pyrimidines as Tpl2 kinase inhibitors
- Synthesis of pyridinyl benzonaphthyridinones as mTOR inhibitors for the treatment of cancer
- Novel bisphosphonate inhibitors of human farnesyl pyrophosphate synthase
- Preparation of diazabicycloheptanes as α7 neurinal nicotinic receptor affectors
- Preparation of pyrazolopyrimidinamine derivatives with tyrosine and phosphinositide kinase inhibitory activity
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service