Skip to Content
Merck
All Photos(2)

Key Documents

684090

Sigma-Aldrich

2-Aminopyridine-3-carboxamide

97%

Synonym(s):

2-Aminonicotinamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N3O
CAS Number:
Molecular Weight:
137.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

mp

194-200 °C

SMILES string

NC(=O)c1cccnc1N

InChI

1S/C6H7N3O/c7-5-4(6(8)10)2-1-3-9-5/h1-3H,(H2,7,9)(H2,8,10)

InChI key

HTPCDVLWYUXWQR-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher L Frank et al.
Toxicological sciences : an official journal of the Society of Toxicology, 160(1), 121-135 (2017-10-04)
Less than 1% of environmental chemicals have been evaluated for developmental neurotoxicity (DNT). Current guideline DNT studies are resource intensive and not amenable to screening large numbers of compounds for hazard. As part of evaluating a battery of more rapid
Haizhen Wang et al.
Nature, 546(7658), 426-430 (2017-06-14)
D-type cyclins (D1, D2 and D3) and their associated cyclin-dependent kinases (CDK4 and CDK6) are components of the core cell cycle machinery that drives cell proliferation. Inhibitors of CDK4 and CDK6 are currently being tested in clinical trials for patients
Joshua A Harrill et al.
Toxicology and applied pharmacology, 354, 24-39 (2018-04-08)
Medium- to high-throughput in vitro assays that recapitulate the critical processes of nervous system development have been proposed as a means to facilitate rapid testing and identification of chemicals which may affect brain development. In vivo neurodevelopment is a complex
Wen-jing Xiao et al.
Acta pharmacologica Sinica, 36(2), 259-267 (2015-01-27)
The pentose phosphate pathway (PPP) is involved in the activity of glucose-6-phosphate dehydrogenase (G6PD) and generation of NADPH, which plays a key role in drug metabolism. The aim of this study was to investigate the effects of modulation of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service