Skip to Content
MilliporeSigma

Skip To

525928

Quinaldoyl chloride

97%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
2 g

Available to ship TODAYfromMILWAUKEE

$193.00

About This Item

Empirical Formula (Hill Notation):
C10H6ClNO
CAS Number:
Molecular Weight:
191.61
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
MDL number:
Assay:
97%

$193.00


Available to ship TODAYDetails


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

97%

mp

96-98 °C (dec.) (lit.)

functional group

acyl chloride

storage temp.

2-8°C

SMILES string

ClC(=O)c1ccc2ccccc2n1

InChI

1S/C10H6ClNO/c11-10(13)9-6-5-7-3-1-2-4-8(7)12-9/h1-6H

InChI key

WFVMVMAUXYOQSW-UHFFFAOYSA-N

General description

Quinaldoyl chloride is a quinaldine derivative.

Application

Quinaldoyl chloride may be used to synthesize 5-chloro-2-(2-quinolinecarboxy)acetophenone[1] and benzoin quinaldate.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
545708524832384291
assay

97%

assay

97%

assay

96%

assay

95%

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

100

mp

96-98 °C (dec.) (lit.)

mp

53-56 °C (lit.)

mp

147-151 °C (lit.)

mp

45-50 °C (lit.)

storage temp.

2-8°C

storage temp.

-

storage temp.

-

storage temp.

-

functional group

acyl chloride

functional group

chloro

functional group

chloro

functional group

amine, chloro


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library