Skip to Content
Merck
All Photos(1)

Key Documents

521183

Sigma-Aldrich

2-Ethynyl-α,α,α-trifluorotoluene

97%

Synonym(s):

1-Ethynyl-2-trifluoromethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CC6H4CF3
CAS Number:
Molecular Weight:
170.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.4760 (lit.)

bp

45 °C/8 mmHg (lit.)

density

1.222 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1ccccc1C#C

InChI

1S/C9H5F3/c1-2-7-5-3-4-6-8(7)9(10,11)12/h1,3-6H

InChI key

OOZKONVIIMFOKW-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Emma van Dijkum et al.
Organic & biomolecular chemistry, 7(6), 1097-1105 (2009-03-06)
A series of D-glucose derivatives that have been modified at C-4 were synthesised from D-galactose as potential chain terminators of cellulose biosynthesis. Two compounds displayed herbicidal activity in pre-emergence tests and in addition a cell expansion assay at higher concentrations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service