Skip to Content
Merck
All Photos(2)

Documents

476226

Sigma-Aldrich

2-(2-Ethylhexyloxy)ethanol

reagent grade, 97%

Synonym(s):

Ethylene glycol 2-ethylhexyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(C2H5)CH2OCH2CH2OH
CAS Number:
Molecular Weight:
174.28
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

reagent grade

Assay

97%

refractive index

n20/D 1.435 (lit.)

bp

229 °C (lit.)

density

0.892 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)COCCO

InChI

1S/C10H22O2/c1-3-5-6-10(4-2)9-12-8-7-11/h10-11H,3-9H2,1-2H3

InChI key

OHJYHAOODFPJOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service